Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATTTCCTGGATGTGCGCATCCTT[C/T]CAGAGGTTGGGGGTCTGGAAGGTCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608702 | |||||||||||||||||||||||
Literature Links: |
LOC100507291 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LOC100507291 - uncharacterized LOC100507291 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NMNAT3 - nicotinamide nucleotide adenylyltransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001200047.2 | 941 | Nonsense Mutation | TGA,TGG | *,W 59 | NP_001186976.1 | |
NM_001320510.1 | 941 | Nonsense Mutation | TGA,TGG | *,W 242 | NP_001307439.1 | |
NM_001320511.1 | 941 | Nonsense Mutation | TGA,TGG | *,W 148 | NP_001307440.1 | |
NM_001320512.1 | 941 | Nonsense Mutation | TGA,TGG | *,W 148 | NP_001307441.1 | |
NM_001320513.1 | 941 | Nonsense Mutation | TGA,TGG | *,W 38 | NP_001307442.1 | |
NM_178177.4 | 941 | Nonsense Mutation | TGA,TGG | *,W 111 | NP_835471.1 | |
XM_011512790.1 | 941 | Nonsense Mutation | TGA,TGG | *,W 111 | XP_011511092.1 | |
XM_011512791.2 | 941 | Nonsense Mutation | TGA,TGG | *,W 111 | XP_011511093.1 | |
XM_017006328.1 | 941 | Nonsense Mutation | TGA,TGG | *,W 242 | XP_016861817.1 | |
XM_017006329.1 | 941 | Nonsense Mutation | TGA,TGG | *,W 242 | XP_016861818.1 | |
XM_017006330.1 | 941 | Nonsense Mutation | TGA,TGG | *,W 242 | XP_016861819.1 | |
XM_017006331.1 | 941 | Nonsense Mutation | TGA,TGG | *,W 242 | XP_016861820.1 | |
XM_017006332.1 | 941 | Nonsense Mutation | TGA,TGG | *,W 242 | XP_016861821.1 | |
XM_017006333.1 | 941 | Nonsense Mutation | TGA,TGG | *,W 242 | XP_016861822.1 | |
XM_017006334.1 | 941 | Nonsense Mutation | TGA,TGG | *,W 242 | XP_016861823.1 | |
XM_017006335.1 | 941 | Nonsense Mutation | TGA,TGG | *,W 148 | XP_016861824.1 | |
XM_017006336.1 | 941 | Nonsense Mutation | TGA,TGG | *,W 148 | XP_016861825.1 | |
XM_017006337.1 | 941 | Nonsense Mutation | TGA,TGG | *,W 148 | XP_016861826.1 | |
XM_017006338.1 | 941 | Nonsense Mutation | TGA,TGG | *,W 111 | XP_016861827.1 | |
XM_017006339.1 | 941 | UTR 3 | XP_016861828.1 | |||
XM_017006340.1 | 941 | UTR 3 | XP_016861829.1 | |||
XM_017006341.1 | 941 | UTR 3 | XP_016861830.1 | |||
XM_017006342.1 | 941 | UTR 3 | XP_016861831.1 | |||
XM_017006343.1 | 941 | UTR 3 | XP_016861832.1 | |||
XM_017006344.1 | 941 | UTR 3 | XP_016861833.1 |