Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGCCACCACCTATGGAGCGCAC[A/G]GGAGGCATCGGGGACTCCCGACCCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601368 | |||||||||||||||||||||||
Literature Links: |
DVL3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DVL3 - dishevelled segment polarity protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004423.3 | 643 | Silent Mutation | ACA,ACG | T,T 106 | NP_004414.3 | |
XM_005247172.2 | 643 | Silent Mutation | ACA,ACG | T,T 106 | XP_005247229.1 | |
XM_011512513.2 | 643 | UTR 5 | XP_011510815.1 |