Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGCACAGCCAACATGTGTGACA[C/T]TGAAGAAGCCACTAACCAACTCCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606525 | ||||||||||||||||||||
Literature Links: |
SRGAP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SRGAP3 - SLIT-ROBO Rho GTPase activating protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THUMPD3 - THUMP domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114092.1 | 721 | Missense Mutation | ACT,ATT | T,I 4 | NP_001107564.1 | |
NM_015453.2 | 721 | Missense Mutation | ACT,ATT | T,I 4 | NP_056268.2 | |
XM_005265021.4 | 721 | Missense Mutation | ACT,ATT | T,I 4 | XP_005265078.1 | |
XM_005265022.4 | 721 | Missense Mutation | ACT,ATT | T,I 4 | XP_005265079.1 | |
XM_005265023.3 | 721 | Missense Mutation | ACT,ATT | T,I 4 | XP_005265080.1 | |
XM_005265024.4 | 721 | Missense Mutation | ACT,ATT | T,I 4 | XP_005265081.1 | |
XM_006713086.3 | 721 | Missense Mutation | ACT,ATT | T,I 4 | XP_006713149.1 | |
XM_011533570.2 | 721 | Missense Mutation | ACT,ATT | T,I 4 | XP_011531872.1 | |
XM_011533571.2 | 721 | Missense Mutation | ACT,ATT | T,I 4 | XP_011531873.1 | |
XM_017006113.1 | 721 | Missense Mutation | ACT,ATT | T,I 4 | XP_016861602.1 | |
XM_017006114.1 | 721 | Missense Mutation | ACT,ATT | T,I 4 | XP_016861603.1 | |
XM_017006115.1 | 721 | Missense Mutation | ACT,ATT | T,I 4 | XP_016861604.1 | |
XM_017006116.1 | 721 | Missense Mutation | ACT,ATT | T,I 4 | XP_016861605.1 | |
XM_017006117.1 | 721 | Missense Mutation | ACT,ATT | T,I 4 | XP_016861606.1 | |
XM_017006118.1 | 721 | Missense Mutation | ACT,ATT | T,I 4 | XP_016861607.1 | |
XM_017006119.1 | 721 | Missense Mutation | ACT,ATT | T,I 4 | XP_016861608.1 |