Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCATTAACAAACTTAAACCTTCA[A/G]ATGTAAGTATCTTTGTTAGATGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608845 MIM: 600066 | ||||||||||||||||||||
Literature Links: |
ARL6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARL6 - ADP ribosylation factor like GTPase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006713783.3 | 2287 | Missense Mutation | AAT,GAT | N,D 41 | XP_006713846.1 | |
XM_017007311.1 | 2287 | Missense Mutation | AAT,GAT | N,D 41 | XP_016862800.1 | |
XM_017007312.1 | 2287 | Missense Mutation | AAT,GAT | N,D 41 | XP_016862801.1 | |
XM_017007313.1 | 2287 | Missense Mutation | AAT,GAT | N,D 41 | XP_016862802.1 |
EPHA6 - EPH receptor A6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |