Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAGAAGGTCATCTCCACTTACG[A/T]ATCGGAGGATTGGAATCTCCTCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ERICH6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ERICH6 - glutamate rich 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308234.1 | 2199 | Missense Mutation | TAC,TTC | Y,F 442 | NP_001295163.1 | |
NM_152394.4 | 2199 | Missense Mutation | TAC,TTC | Y,F 588 | NP_689607.2 | |
XM_005247120.3 | 2199 | Missense Mutation | ACG,TCG | T,S 512 | XP_005247177.1 |