Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCAGGCGGCAGCTGAAGGAGAG[G/T]TCGGAGCCCTCGGGGTCTGCGCCGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608721 MIM: 610145 | |||||||||||||||||||||||
Literature Links: |
CAMK2N2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CAMK2N2 - calcium/calmodulin dependent protein kinase II inhibitor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033259.2 | 247 | Missense Mutation | GAA,GAC | E,D 23 | NP_150284.1 |
ECE2 - endothelin converting enzyme 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037324.2 | 247 | Intron | NP_001032401.1 | |||
NM_001100120.1 | 247 | Intron | NP_001093590.1 | |||
NM_001100121.1 | 247 | Intron | NP_001093591.1 | |||
NM_014693.3 | 247 | Intron | NP_055508.3 | |||
NM_032331.3 | 247 | Intron | NP_115707.2 |