Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGACTGGAATAAGCAAAACTTG[C/T]CCGGGCAGTCAGATCCATTTCTCCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 150210 | |||||||||||||||||||||||
Literature Links: |
LTF PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LTF - lactotransferrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199149.1 | 2128 | Missense Mutation | GAC,GGC | D,G 604 | NP_001186078.1 | |
NM_001321121.1 | 2128 | Missense Mutation | GAC,GGC | D,G 646 | NP_001308050.1 | |
NM_001321122.1 | 2128 | Missense Mutation | GAC,GGC | D,G 635 | NP_001308051.1 | |
NM_002343.5 | 2128 | Missense Mutation | GAC,GGC | D,G 648 | NP_002334.2 |