Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 146691 MIM: 615150 MIM: 612911 MIM: 614584 MIM: 606031 | |||||||||||||||||||||||
Literature Links: |
DALRD3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DALRD3 - DALR anticodon binding domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009996.2 | 1351 | Silent Mutation | ACC,ACT | T,T 325 | NP_001009996.1 | |
NM_001276405.1 | 1351 | Silent Mutation | ACC,ACT | T,T 325 | NP_001263334.1 | |
NM_018114.5 | 1351 | Silent Mutation | ACC,ACT | T,T 158 | NP_060584.3 | |
XM_005265269.3 | 1351 | Silent Mutation | ACC,ACT | T,T 325 | XP_005265326.1 | |
XM_006713219.2 | 1351 | Silent Mutation | ACC,ACT | T,T 158 | XP_006713282.1 | |
XM_011533891.1 | 1351 | Silent Mutation | ACC,ACT | T,T 325 | XP_011532193.1 | |
XM_011533892.1 | 1351 | Silent Mutation | ACC,ACT | T,T 158 | XP_011532194.1 | |
XM_011533893.1 | 1351 | Silent Mutation | ACC,ACT | T,T 325 | XP_011532195.1 | |
XM_017006723.1 | 1351 | Silent Mutation | ACC,ACT | T,T 158 | XP_016862212.1 |
IMPDH2 - inosine monophosphate dehydrogenase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR191 - microRNA 191 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR425 - microRNA 425 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFAF3 - NADH:ubiquinone oxidoreductase complex assembly factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
P4HTM - prolyl 4-hydroxylase, transmembrane | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR6 - WD repeat domain 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320546.1 | 1351 | Intron | NP_001307475.1 | |||
NM_001320547.1 | 1351 | Intron | NP_001307476.1 | |||
NM_018031.4 | 1351 | Intron | NP_060501.3 |