Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTCCTTCCCCTGGCACTCTGCA[A/T]CCTAGTCCTGTGCTGCGGAGCAATG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605954 | |||||||||||||||||||||||
Literature Links: |
FETUB PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
FETUB - fetuin B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308077.1 | 133 | Missense Mutation | AAC,ATC | N,I 11 | NP_001295006.1 | |
NM_001308079.1 | 133 | Missense Mutation | AAC,ATC | N,I 11 | NP_001295008.1 | |
NM_014375.2 | 133 | Missense Mutation | AAC,ATC | N,I 11 | NP_055190.2 | |
XM_005247351.3 | 133 | UTR 5 | XP_005247408.1 | |||
XM_011512681.2 | 133 | Missense Mutation | AAC,ATC | N,I 11 | XP_011510983.1 | |
XM_011512682.2 | 133 | UTR 5 | XP_011510984.1 | |||
XM_011512683.2 | 133 | Intron | XP_011510985.1 |