Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCAAGCTGCCTGGAGTCAAGGA[A/G]GGTAAGGGCCCCGGAGGGAGAGCGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 611051 | |||||||||||||||||||||||
Literature Links: |
CCDC50 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CCDC50 - coiled-coil domain containing 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174908.3 | 641 | Silent Mutation | GAA,GAG | E,E 16 | NP_777568.1 | |
NM_178335.2 | 641 | Silent Mutation | GAA,GAG | E,E 16 | NP_848018.1 | |
XM_011512460.1 | 641 | Silent Mutation | GAA,GAG | E,E 16 | XP_011510762.1 |
UTS2B - urotensin 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198152.3 | 641 | Intron | NP_937795.2 | |||
XM_011512631.1 | 641 | Intron | XP_011510933.1 | |||
XM_017006091.1 | 641 | Intron | XP_016861580.1 |