Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGTGCTGTCCTCCACGAGTCTCC[A/G]GCTGCACTCCAACCCTGAGCCCAAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
KIF15 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
KIF15 - kinesin family member 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020242.2 | 331 | Missense Mutation | CAG,CGG | Q,R 60 | NP_064627.1 | |
XM_006713264.3 | 331 | Intron | XP_006713327.1 | |||
XM_011533964.2 | 331 | Missense Mutation | CAG,CGG | Q,R 60 | XP_011532266.1 | |
XM_017006884.1 | 331 | Missense Mutation | CAG,CGG | Q,R 60 | XP_016862373.1 | |
XM_017006885.1 | 331 | Missense Mutation | AGC,GGC | S,G 47 | XP_016862374.1 | |
XM_017006886.1 | 331 | UTR 5 | XP_016862375.1 | |||
XM_017006887.1 | 331 | Missense Mutation | CAG,CGG | Q,R 60 | XP_016862376.1 | |
XM_017006888.1 | 331 | UTR 5 | XP_016862377.1 | |||
XM_017006889.1 | 331 | Missense Mutation | CAG,CGG | Q,R 60 | XP_016862378.1 | |
XM_017006890.1 | 331 | Intron | XP_016862379.1 |