Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAGCAGTGAAAAGGAAGAAGAC[A/G]ATAATGAAAAGAGACAAGATCCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CDV3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDV3 - CDV3 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134422.1 | 816 | Missense Mutation | AAT,GAT | N,D 113 | NP_001127894.1 | |
NM_001134423.2 | 816 | Missense Mutation | AAT,GAT | N,D 11 | NP_001127895.1 | |
NM_001282762.1 | 816 | Missense Mutation | AAT,GAT | N,D 11 | NP_001269691.1 | |
NM_001282763.1 | 816 | Missense Mutation | AAT,GAT | N,D 11 | NP_001269692.1 | |
NM_001282764.1 | 816 | Missense Mutation | AAT,GAT | N,D 11 | NP_001269693.1 | |
NM_001282765.1 | 816 | Missense Mutation | AAT,GAT | N,D 11 | NP_001269694.1 | |
NM_017548.4 | 816 | Missense Mutation | AAT,GAT | N,D 113 | NP_060018.1 | |
XM_005247588.4 | 816 | Missense Mutation | AAT,GAT | N,D 113 | XP_005247645.1 | |
XM_005247589.4 | 816 | Missense Mutation | AAT,GAT | N,D 112 | XP_005247646.1 | |
XM_005247590.4 | 816 | Missense Mutation | AAT,GAT | N,D 112 | XP_005247647.1 | |
XM_005247591.4 | 816 | Missense Mutation | AAT,GAT | N,D 113 | XP_005247648.1 | |
XM_005247592.4 | 816 | Missense Mutation | AAT,GAT | N,D 11 | XP_005247649.1 | |
XM_005247594.4 | 816 | Missense Mutation | AAT,GAT | N,D 10 | XP_005247651.1 | |
XM_011512961.2 | 816 | Missense Mutation | AAT,GAT | N,D 113 | XP_011511263.1 | |
XM_011512962.2 | 816 | Missense Mutation | AAT,GAT | N,D 11 | XP_011511264.1 | |
XM_017006808.1 | 816 | Missense Mutation | AAT,GAT | N,D 11 | XP_016862297.1 |