Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACAGTGGTTCTTAAGTACTTAC[A/G]TGGGCAATGTCTCCTTTTGTGCCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DCAF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCAF1 - DDB1 and CUL4 associated factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171904.1 | 3649 | Silent Mutation | CAC,CAT | H,H 1200 | NP_001165375.1 | |
NM_014703.2 | 3649 | Silent Mutation | CAC,CAT | H,H 1201 | NP_055518.1 | |
XM_005276753.4 | 3649 | Silent Mutation | CAC,CAT | H,H 1201 | XP_005276810.1 | |
XM_005276755.4 | 3649 | Silent Mutation | CAC,CAT | H,H 1201 | XP_005276812.1 | |
XM_011534273.2 | 3649 | Silent Mutation | CAC,CAT | H,H 1201 | XP_011532575.1 | |
XM_011534274.2 | 3649 | Silent Mutation | CAC,CAT | H,H 1201 | XP_011532576.1 | |
XM_011534275.2 | 3649 | Silent Mutation | CAC,CAT | H,H 1201 | XP_011532577.1 | |
XM_011534276.2 | 3649 | Silent Mutation | CAC,CAT | H,H 1201 | XP_011532578.1 | |
XM_011534277.2 | 3649 | Silent Mutation | CAC,CAT | H,H 1201 | XP_011532579.1 | |
XM_017007546.1 | 3649 | Silent Mutation | CAC,CAT | H,H 1201 | XP_016863035.1 | |
XM_017007547.1 | 3649 | Silent Mutation | CAC,CAT | H,H 1201 | XP_016863036.1 | |
XM_017007548.1 | 3649 | Silent Mutation | CAC,CAT | H,H 1201 | XP_016863037.1 | |
XM_017007549.1 | 3649 | Silent Mutation | CAC,CAT | H,H 1201 | XP_016863038.1 | |
XM_017007550.1 | 3649 | Silent Mutation | CAC,CAT | H,H 1201 | XP_016863039.1 | |
XM_017007551.1 | 3649 | Silent Mutation | CAC,CAT | H,H 1119 | XP_016863040.1 |