Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTGCGACCTGCTGCAGGCCATGC[C/T]CCGGACCGAGTTCGCGCGCCTCAAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 612200 | |||||||||||||||||||||||
Literature Links: |
C3orf58 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
C3orf58 - chromosome 3 open reading frame 58 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134470.1 | 960 | Intron | NP_001127942.1 | |||
NM_173552.3 | 960 | Missense Mutation | CCC,CTC | P,L 142 | NP_775823.1 | |
XM_011512543.2 | 960 | Missense Mutation | CCC,CTC | P,L 142 | XP_011510845.1 | |
XM_017005869.1 | 960 | Intron | XP_016861358.1 |