Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAATTGGTATAGAAAATCTTCAAT[C/T]TGTCCAACCTAGAAGAAATAAACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609856 | ||||||||||||||||||||
Literature Links: |
SPATA16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPATA16 - spermatogenesis associated 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031955.5 | 1780 | Silent Mutation | CAA,CAG | Q,Q 532 | NP_114161.3 | |
XM_006713778.2 | 1780 | Intron | XP_006713841.1 | |||
XM_017007308.1 | 1780 | Intron | XP_016862797.1 |