Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGAAGCTGTAAGTCCATCCGGTC[C/T]GTCAACCAGGTGCAGATCACGTTCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604667 | |||||||||||||||||||||||
Literature Links: |
CADPS PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CADPS - calcium dependent secretion activator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003716.3 | 4363 | Silent Mutation | ACA,ACG | T,T 1274 | NP_003707.2 | |
NM_183393.2 | 4363 | Silent Mutation | ACA,ACG | T,T 1195 | NP_899630.1 | |
NM_183394.2 | 4363 | Silent Mutation | ACA,ACG | T,T 1235 | NP_899631.1 | |
XM_006713378.2 | 4363 | Silent Mutation | ACA,ACG | T,T 1216 | XP_006713441.2 | |
XM_011534177.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1352 | XP_011532479.1 | |
XM_011534178.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1346 | XP_011532480.1 | |
XM_011534180.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1336 | XP_011532482.1 | |
XM_011534182.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1329 | XP_011532484.1 | |
XM_011534183.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1323 | XP_011532485.1 | |
XM_011534189.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1297 | XP_011532491.1 | |
XM_011534191.2 | 4363 | Silent Mutation | ACA,ACG | T,T 1284 | XP_011532493.1 | |
XM_011534192.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1283 | XP_011532494.1 | |
XM_011534193.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1279 | XP_011532495.1 | |
XM_011534194.2 | 4363 | Silent Mutation | ACA,ACG | T,T 1278 | XP_011532496.1 | |
XM_011534195.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1274 | XP_011532497.1 | |
XM_011534196.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1273 | XP_011532498.1 | |
XM_011534197.2 | 4363 | Silent Mutation | ACA,ACG | T,T 1265 | XP_011532499.1 | |
XM_011534198.2 | 4363 | Silent Mutation | ACA,ACG | T,T 1218 | XP_011532500.1 | |
XM_011534199.2 | 4363 | Silent Mutation | ACA,ACG | T,T 1217 | XP_011532501.1 | |
XM_011534200.2 | 4363 | Silent Mutation | ACA,ACG | T,T 1212 | XP_011532502.1 | |
XM_011534202.2 | 4363 | Intron | XP_011532504.1 | |||
XM_011534203.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1268 | XP_011532505.1 | |
XM_017007358.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1354 | XP_016862847.1 | |
XM_017007359.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1350 | XP_016862848.1 | |
XM_017007360.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1347 | XP_016862849.1 | |
XM_017007361.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1331 | XP_016862850.1 | |
XM_017007362.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1327 | XP_016862851.1 | |
XM_017007363.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1324 | XP_016862852.1 | |
XM_017007364.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1298 | XP_016862853.1 | |
XM_017007365.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1294 | XP_016862854.1 | |
XM_017007366.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1292 | XP_016862855.1 | |
XM_017007367.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1290 | XP_016862856.1 | |
XM_017007368.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1289 | XP_016862857.1 | |
XM_017007369.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1283 | XP_016862858.1 | |
XM_017007370.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1278 | XP_016862859.1 | |
XM_017007371.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1275 | XP_016862860.1 | |
XM_017007372.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1275 | XP_016862861.1 | |
XM_017007373.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1274 | XP_016862862.1 | |
XM_017007374.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1271 | XP_016862863.1 | |
XM_017007375.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1270 | XP_016862864.1 | |
XM_017007376.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1269 | XP_016862865.1 | |
XM_017007377.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1267 | XP_016862866.1 | |
XM_017007378.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1266 | XP_016862867.1 | |
XM_017007379.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1265 | XP_016862868.1 | |
XM_017007380.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1246 | XP_016862869.1 | |
XM_017007381.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1245 | XP_016862870.1 | |
XM_017007382.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1241 | XP_016862871.1 | |
XM_017007383.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1239 | XP_016862872.1 | |
XM_017007384.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1229 | XP_016862873.1 | |
XM_017007385.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1225 | XP_016862874.1 | |
XM_017007386.1 | 4363 | Silent Mutation | ACA,ACG | T,T 1222 | XP_016862875.1 |