Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGACCAATTTCTTCAAACTACGG[C/T]TTTTCTGCTGTCTGCTTGCAGTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
NXPE3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NXPE3 - neurexophilin and PC-esterase domain family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134456.1 | 665 | Missense Mutation | CTT,TTT | L,F 10 | NP_001127928.1 | |
NM_145037.2 | 665 | Missense Mutation | CTT,TTT | L,F 10 | NP_659474.1 |