Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGTCATGATGTCCCACGGGCAG[A/G]GACTCTGGGCCCCTTCGAGGGGTGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 616613 MIM: 602610 | |||||||||||||||||||||||
Literature Links: |
COL6A6 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
COL6A6 - collagen type VI alpha 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIK3R4 - phosphoinositide-3-kinase regulatory subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014602.2 | 4550 | Silent Mutation | TCC,TCT | S,S 1323 | NP_055417.1 |