Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACAGACTTACAATGAGGAAACA[A/G]GCACCAATCATTACAGCTTTCATTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604170 | |||||||||||||||||||||||
Literature Links: |
PLSCR1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PLSCR1 - phospholipid scramblase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021105.2 | 996 | Silent Mutation | GCC,GCT | A,A 296 | NP_066928.1 | |
XM_005247538.3 | 996 | Silent Mutation | GCC,GCT | A,A 215 | XP_005247595.1 | |
XM_011512904.1 | 996 | Silent Mutation | GCC,GCT | A,A 296 | XP_011511206.1 | |
XM_011512905.1 | 996 | Silent Mutation | GCC,GCT | A,A 289 | XP_011511207.1 | |
XM_011512906.2 | 996 | Silent Mutation | GCC,GCT | A,A 215 | XP_011511208.1 | |
XM_011512907.2 | 996 | Silent Mutation | GCC,GCT | A,A 215 | XP_011511209.1 | |
XM_017006628.1 | 996 | Silent Mutation | GCC,GCT | A,A 296 | XP_016862117.1 | |
XM_017006629.1 | 996 | Silent Mutation | GCC,GCT | A,A 296 | XP_016862118.1 |