Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACGAAGGAAGCTGGAGAGCTACTT[A/C]CAGAGCTCTAAGTCCTCGGGCGGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613143 MIM: 612065 | ||||||||||||||||||||
Literature Links: |
DTX3L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DTX3L - deltex E3 ubiquitin ligase 3L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138287.3 | 179 | Silent Mutation | TTA,TTC | L,F 30 | NP_612144.1 |
PARP9 - poly(ADP-ribose) polymerase family member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146102.1 | 179 | Intron | NP_001139574.1 | |||
NM_001146103.1 | 179 | UTR 5 | NP_001139575.1 | |||
NM_001146104.1 | 179 | Intron | NP_001139576.1 | |||
NM_001146105.1 | 179 | Intron | NP_001139577.1 | |||
NM_001146106.1 | 179 | Intron | NP_001139578.1 | |||
NM_031458.2 | 179 | UTR 5 | NP_113646.2 | |||
XM_005247820.2 | 179 | Intron | XP_005247877.1 | |||
XM_011513218.1 | 179 | Intron | XP_011511520.1 | |||
XM_017007304.1 | 179 | Intron | XP_016862793.1 | |||
XM_017007305.1 | 179 | Intron | XP_016862794.1 | |||
XM_017007306.1 | 179 | Intron | XP_016862795.1 |