Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGGATATCTCTTCCAAATGCAT[A/G]ATGAAGAAGTTCTCATCAACAGCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606738 | ||||||||||||||||||||
Literature Links: |
OSBPL10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OSBPL10 - oxysterol binding protein like 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF860 - zinc finger protein 860 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137674.2 | 759 | Missense Mutation | ATA,ATG | I,M 70 | NP_001131146.2 | |
XM_017006299.1 | 759 | Intron | XP_016861788.1 |