Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGGAGGTGCTGAAGACCAATTA[C/T]GGCATCCCCTCCGCCTGCTTCTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612084 | ||||||||||||||||||||
Literature Links: |
SLC51A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC51A - solute carrier family 51 alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152672.5 | 279 | Silent Mutation | TAC,TAT | Y,Y 26 | NP_689885.4 |
ZDHHC19 - zinc finger DHHC-type containing 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |