Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACTGGAATAAGCAAAACTTGTCC[A/G]GGCAGTCAGATCCATTTCTCCCAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 150210 | ||||||||||||||||||||
Literature Links: |
LTF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LTF - lactotransferrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199149.1 | 2125 | Missense Mutation | CCG,CTG | P,L 603 | NP_001186078.1 | |
NM_001321121.1 | 2125 | Missense Mutation | CCG,CTG | P,L 645 | NP_001308050.1 | |
NM_001321122.1 | 2125 | Missense Mutation | CCG,CTG | P,L 634 | NP_001308051.1 | |
NM_002343.5 | 2125 | Missense Mutation | CCG,CTG | P,L 647 | NP_002334.2 |