Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCATGACTTTTCAAAGCACAGTT[C/T]CTAGAGAGTTTTCACACGCTGTCAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 616699 | |||||||||||||||||||||||
Literature Links: |
ANKUB1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ANKUB1 - ankyrin repeat and ubiquitin domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144960.2 | 1576 | Missense Mutation | GAA,GGA | E,G 541 | NP_001138432.1 | |
NM_001315505.1 | 1576 | Missense Mutation | GAA,GGA | E,G 395 | NP_001302434.1 | |
NM_001315506.1 | 1576 | Intron | NP_001302435.1 | |||
XM_011512798.1 | 1576 | Missense Mutation | GAA,GGA | E,G 492 | XP_011511100.1 | |
XM_011512799.1 | 1576 | Intron | XP_011511101.1 | |||
XM_011512801.1 | 1576 | Missense Mutation | GAA,GGA | E,G 375 | XP_011511103.1 | |
XM_011512802.1 | 1576 | Intron | XP_011511104.1 |
COMMD2 - COMM domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |