Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGTGGTGAGGAGGCTGTGGCTG[G/T]GGTGGAGGATTTCCAGAGACACCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZKSCAN7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZKSCAN7 - zinc finger with KRAB and SCAN domains 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288590.1 | 808 | Missense Mutation | GGG,GTG | G,V 128 | NP_001275519.1 | |
NM_001288591.1 | 808 | Missense Mutation | GGG,GTG | G,V 128 | NP_001275520.1 | |
NM_001288592.1 | 808 | Intron | NP_001275521.1 | |||
NM_018651.3 | 808 | Missense Mutation | GGG,GTG | G,V 128 | NP_061121.2 | |
NM_025169.2 | 808 | Missense Mutation | GGG,GTG | G,V 128 | NP_079445.1 |