Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGTGGCTGCCTCAGCACCTCGAG[A/G]ATCGACATGGACGCTCTCGAGGACT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614584 MIM: 606031 | |||||||||||||||||||||||
Literature Links: |
DALRD3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DALRD3 - DALR anticodon binding domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
P4HTM - prolyl 4-hydroxylase, transmembrane | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177938.2 | 364 | Intron | NP_808807.2 | |||
NM_177939.2 | 364 | Intron | NP_808808.1 |
WDR6 - WD repeat domain 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320546.1 | 364 | UTR 5 | NP_001307475.1 | |||
NM_001320547.1 | 364 | Intron | NP_001307476.1 | |||
NM_018031.4 | 364 | Silent Mutation | AGA,AGG | R,R 28 | NP_060501.3 |