Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTTTATATACACAGGAACTTTAA[A/C]TCTTGACAGGTAAAGTACACATTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608534 MIM: 606042 MIM: 602322 | ||||||||||||||||||||
Literature Links: |
ACTRT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACTRT3 - actin related protein T3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYNN - myoneurin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185118.1 | 1444 | Missense Mutation | AAT,ACT | N,T 86 | NP_001172047.1 | |
NM_001185119.1 | 1444 | Missense Mutation | AAT,ACT | N,T 86 | NP_001172048.1 | |
NM_018657.4 | 1444 | Missense Mutation | AAT,ACT | N,T 86 | NP_061127.1 | |
XM_005247621.4 | 1444 | Missense Mutation | AAT,ACT | N,T 86 | XP_005247678.1 | |
XM_005247622.3 | 1444 | Intron | XP_005247679.1 | |||
XM_005247624.3 | 1444 | Intron | XP_005247681.1 | |||
XM_011512987.1 | 1444 | Missense Mutation | AAT,ACT | N,T 86 | XP_011511289.1 | |
XM_017006864.1 | 1444 | Missense Mutation | AAT,ACT | N,T 86 | XP_016862353.1 | |
XM_017006865.1 | 1444 | Intron | XP_016862354.1 | |||
XM_017006866.1 | 1444 | Intron | XP_016862355.1 | |||
XM_017006867.1 | 1444 | UTR 5 | XP_016862356.1 | |||
XM_017006868.1 | 1444 | UTR 5 | XP_016862357.1 |
TERC - telomerase RNA component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |