Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCTCTTCTTCCTCCTCGTAGTG[C/T]TCCTCTCGGCCTTTGGGGCGGTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606805 | ||||||||||||||||||||
Literature Links: |
GOLIM4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GOLIM4 - golgi integral membrane protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308155.1 | 2653 | Silent Mutation | GAA,GAG | E,E 643 | NP_001295084.1 | |
NM_014498.4 | 2653 | Silent Mutation | GAA,GAG | E,E 671 | NP_055313.1 | |
XM_005247364.2 | 2653 | Silent Mutation | GAA,GAG | E,E 672 | XP_005247421.1 | |
XM_005247365.2 | 2653 | Silent Mutation | GAA,GAG | E,E 644 | XP_005247422.1 | |
XM_017006183.1 | 2653 | Silent Mutation | GAA,GAG | E,E 525 | XP_016861672.1 |