Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAAGCCCAAGTGGTCCCGGGTGA[C/T]GGGACACACCAGCTGCTTCTCATTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603104 | ||||||||||||||||||||
Literature Links: |
CPN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CPN2 - carboxypeptidase N subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080513.3 | 1429 | Missense Mutation | ATC,GTC | I,V 447 | NP_001073982.3 | |
NM_001291988.1 | 1429 | Missense Mutation | ATC,GTC | I,V 447 | NP_001278917.1 | |
XM_005269280.4 | 1429 | Missense Mutation | ATC,GTC | I,V 447 | XP_005269337.1 |