Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGATGTTGGTGTCGGGCCGGACC[A/G]TCACGGCGTGGGGTGGTATCTGTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 155750 | ||||||||||||||||||||
Literature Links: |
MELTF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MELTF - melanotransferrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005929.5 | 1993 | Missense Mutation | ACG,ATG | T,M 628 | NP_005920.2 | |
NM_033316.3 | 1993 | Intron | NP_201573.1 | |||
XM_006713643.3 | 1993 | Missense Mutation | ACG,ATG | T,M 627 | XP_006713706.2 | |
XM_011512850.2 | 1993 | Missense Mutation | ACG,ATG | T,M 628 | XP_011511152.1 | |
XM_011512851.2 | 1993 | Missense Mutation | ACG,ATG | T,M 628 | XP_011511153.1 |
MFI2-AS1 - MFI2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |