Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGTGTCAGAGATGGACTGCCACTT[A/T]CTGCTTCTACTGATTATGAACAAAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 612442 | |||||||||||||||||||||||
Literature Links: |
SEC22A PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SEC22A - SEC22 homolog A, vesicle trafficking protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012430.4 | 190 | Missense Mutation | ACT,TCT | T,S 19 | NP_036562.2 | |
XM_011512673.2 | 190 | Missense Mutation | ACT,TCT | T,S 19 | XP_011510975.1 | |
XM_011512675.2 | 190 | Missense Mutation | ACT,TCT | T,S 19 | XP_011510977.1 | |
XM_011512676.2 | 190 | Missense Mutation | ACT,TCT | T,S 19 | XP_011510978.1 |