Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGCGCCGAGGTGCCGGAACCCGC[C/T]GCCCCCAGCAGACTTCACATGTCCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 184429 MIM: 616338 | |||||||||||||||||||||||
Literature Links: |
SOX2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SOX2 - SRY-box 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003106.3 | 1298 | Silent Mutation | GCC,GCT | A,A 287 | NP_003097.1 |
SOX2-OT - SOX2 overlapping transcript | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |