Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAACTTCTGAGTAATGATGAAGT[A/C]ACAATAAAGTATAAAGATGAAGGTA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 602498 | |||||||||||||||||||||||
Literature Links: |
TFG PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TFG - TRK-fused gene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007565.2 | 596 | Silent Mutation | GTA,GTC | V,V 54 | NP_001007566.1 | |
NM_001195478.1 | 596 | Silent Mutation | GTA,GTC | V,V 54 | NP_001182407.1 | |
NM_001195479.1 | 596 | Silent Mutation | GTA,GTC | V,V 54 | NP_001182408.1 | |
NM_006070.5 | 596 | Silent Mutation | GTA,GTC | V,V 54 | NP_006061.2 | |
XM_005247066.1 | 596 | Silent Mutation | GTA,GTC | V,V 54 | XP_005247123.1 | |
XM_006713472.1 | 596 | Silent Mutation | GTA,GTC | V,V 54 | XP_006713535.1 | |
XM_006713473.1 | 596 | Silent Mutation | GTA,GTC | V,V 54 | XP_006713536.1 | |
XM_011512334.1 | 596 | Silent Mutation | GTA,GTC | V,V 54 | XP_011510636.1 | |
XM_017005527.1 | 596 | Silent Mutation | GTA,GTC | V,V 54 | XP_016861016.1 | |
XM_017005528.1 | 596 | Silent Mutation | GTA,GTC | V,V 54 | XP_016861017.1 | |
XM_017005529.1 | 596 | Silent Mutation | GTA,GTC | V,V 54 | XP_016861018.1 | |
XM_017005530.1 | 596 | Silent Mutation | GTA,GTC | V,V 54 | XP_016861019.1 |