Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCTAAGCCCAGGCAAGGCAAGT[C/G]AAGCCAAGACCTGCAGGCGGAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614572 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFP42 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFP42 - ZFP42 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304358.1 | 337 | Missense Mutation | TCA,TGA | S,* 32 | NP_001291287.1 | |
NM_174900.4 | 337 | Intron | NP_777560.2 | |||
XM_011531604.2 | 337 | Nonsense Mutation | TCA,TGA | S,* 32 | XP_011529906.1 | |
XM_011531605.1 | 337 | Nonsense Mutation | TCA,TGA | S,* 32 | XP_011529907.1 | |
XM_011531606.1 | 337 | Nonsense Mutation | TCA,TGA | S,* 32 | XP_011529908.1 | |
XM_011531607.2 | 337 | Nonsense Mutation | TCA,TGA | S,* 32 | XP_011529909.1 |