Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGGCCGCTTTCCCCATTAATCA[A/G]CATCTTCTCCTGCTGTTCTGCAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603310 | ||||||||||||||||||||
Literature Links: |
LOC645513 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC645513 - septin 7 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDE5A - phosphodiesterase 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083.3 | 2576 | Silent Mutation | CTG,TTG | L,L 864 | NP_001074.2 | |
NM_033430.2 | 2576 | Silent Mutation | CTG,TTG | L,L 822 | NP_236914.2 | |
NM_033437.3 | 2576 | Silent Mutation | CTG,TTG | L,L 812 | NP_246273.2 | |
XM_017008791.1 | 2576 | Intron | XP_016864280.1 |