Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTCACTGGCGTGGTCTCTATCC[A/G]CCTCGGGCTTGACGGTTTGGCCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602618 MIM: 605918 | ||||||||||||||||||||
Literature Links: |
CTBP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTBP1 - C-terminal binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012614.1 | 7403 | Missense Mutation | GCG,GTG | A,V 420 | NP_001012632.1 | |
NM_001328.2 | 7403 | Missense Mutation | GCG,GTG | A,V 431 | NP_001319.1 | |
XM_005272261.3 | 7403 | Missense Mutation | GCG,GTG | A,V 432 | XP_005272318.1 | |
XM_005272263.4 | 7403 | Missense Mutation | GCG,GTG | A,V 421 | XP_005272320.1 | |
XM_011513395.2 | 7403 | Missense Mutation | GCG,GTG | A,V 421 | XP_011511697.1 | |
XM_011513396.2 | 7403 | Missense Mutation | GCG,GTG | A,V 421 | XP_011511698.1 | |
XM_011513397.2 | 7403 | Missense Mutation | GCG,GTG | A,V 358 | XP_011511699.1 | |
XM_017007762.1 | 7403 | Missense Mutation | GCG,GTG | A,V 421 | XP_016863251.1 | |
XM_017007763.1 | 7403 | Missense Mutation | GCG,GTG | A,V 421 | XP_016863252.1 | |
XM_017007764.1 | 7403 | Missense Mutation | GCG,GTG | A,V 420 | XP_016863253.1 | |
XM_017007765.1 | 7403 | Missense Mutation | GCG,GTG | A,V 420 | XP_016863254.1 | |
XM_017007766.1 | 7403 | Missense Mutation | GCG,GTG | A,V 420 | XP_016863255.1 | |
XM_017007767.1 | 7403 | Missense Mutation | GCG,GTG | A,V 420 | XP_016863256.1 |
CTBP1-AS - CTBP1 antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100130872 - uncharacterized LOC100130872 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPON2 - spondin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |