Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCTGGGCGGCATGCGATGGGGAA[A/C]TGCTGCTGGACGCAGTGCTTCGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610851 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP1AR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP1AR - adaptor related protein complex 1 associated regulatory protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128426.2 | 362 | Missense Mutation | AAA,AAC | K,N 3 | NP_001121898.1 | |
NM_018569.5 | 362 | Missense Mutation | AAA,AAC | K,N 3 | NP_061039.3 | |
XM_017008378.1 | 362 | UTR 5 | XP_016863867.1 | |||
XM_017008379.1 | 362 | UTR 5 | XP_016863868.1 | |||
XM_017008380.1 | 362 | Intron | XP_016863869.1 |
LOC105377371 - uncharacterized LOC105377371 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |