Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTGAAATACATTGTGGTCTAAA[C/T]TCTTTGACGAATAAGTAAGGAGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LRP2BP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRP2BP - LRP2 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018409.3 | 1104 | Missense Mutation | ATT,GTT | I,V 347 | NP_060879.2 | |
XM_005263125.4 | 1104 | Missense Mutation | ATT,GTT | I,V 249 | XP_005263182.1 | |
XM_006714260.3 | 1104 | Missense Mutation | ATT,GTT | I,V 347 | XP_006714323.1 | |
XM_006714265.3 | 1104 | Missense Mutation | ATT,GTT | I,V 249 | XP_006714328.1 | |
XM_011532110.2 | 1104 | Missense Mutation | ATT,GTT | I,V 314 | XP_011530412.1 | |
XM_011532111.2 | 1104 | Missense Mutation | ATT,GTT | I,V 249 | XP_011530413.1 | |
XM_017008410.1 | 1104 | Missense Mutation | ATT,GTT | I,V 347 | XP_016863899.1 | |
XM_017008411.1 | 1104 | Missense Mutation | ATT,GTT | I,V 346 | XP_016863900.1 |
SNX25 - sorting nexin 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317781.1 | 1104 | Intron | NP_001304710.1 |