Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCTGACACAGTGAATAAGGTGG[C/T]ACAATGGCTCGATCTCCCTTTTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611744 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTUD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OTUD4 - OTU deubiquitinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102653.1 | Intron | NP_001096123.1 | ||||
NM_017493.6 | Intron | NP_059963.1 | ||||
XM_005263079.4 | Intron | XP_005263136.1 | ||||
XM_011532040.2 | Intron | XP_011530342.2 | ||||
XM_011532041.2 | Intron | XP_011530343.2 | ||||
XM_011532042.2 | Intron | XP_011530344.1 |