Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCCTACTGCTGCAGCTCCTGCC[G/T]CCCGGCCTGCTCCCGGGCTGCGCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611715 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRD5A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SRD5A3 - steroid 5 alpha-reductase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024592.4 | 224 | Silent Mutation | CCG,CCT | P,P 36 | NP_078868.1 | |
XM_005265766.3 | 224 | Silent Mutation | CCG,CCT | P,P 36 | XP_005265823.1 | |
XM_005265767.3 | 224 | Silent Mutation | CCG,CCT | P,P 36 | XP_005265824.1 | |
XM_017008601.1 | 224 | Intron | XP_016864090.1 |