Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGAATCAGGAGGTACCCCTACAA[A/T]GACACTTGTGCCACATGCCTCATGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 103730 | ||||||||||||||||||||
Literature Links: |
ADH1C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADH1C - alcohol dehydrogenase 1C (class I), gamma polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000669.4 | 1034 | Missense Mutation | ATT,TTT | I,F 292 | NP_000660.1 | |
XM_011531588.2 | 1034 | Missense Mutation | ATT,TTT | I,F 258 | XP_011529890.1 | |
XM_011531589.1 | 1034 | Missense Mutation | ATT,TTT | I,F 252 | XP_011529891.1 |