Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTTACCTGAAGGAACATAGAAC[A/G]AATCCCCAGTACTTAATATATAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 117141 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CENPC - centromere protein C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001812.2 | 2927 | Missense Mutation | TCG,TTG | S,L 915 | NP_001803.2 | |
XM_006714064.2 | 2927 | Missense Mutation | TCG,TTG | S,L 907 | XP_006714127.1 | |
XM_011531542.2 | 2927 | Intron | XP_011529844.1 |