Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAACCACCTCCAGAACCCGGGCGA[C/T]GCGGCCGAGGGCCGGGCGGCCAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616905 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRFAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRFAP1 - Morf4 family associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272053.1 | 1033 | Silent Mutation | GAC,GAT | D,D 87 | NP_001258982.1 | |
NM_001272054.1 | 1033 | Silent Mutation | GAC,GAT | D,D 87 | NP_001258983.1 | |
NM_033296.2 | 1033 | Silent Mutation | GAC,GAT | D,D 87 | NP_150638.1 |