Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGAAACACAGAACTTACGGCTG[G/T]GTCATCCTCTCTGTATACTTTAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605961 | ||||||||||||||||||||
Literature Links: |
PLRG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLRG1 - pleiotropic regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201564.1 | 1578 | Missense Mutation | ACA,CCA | T,P 485 | NP_001188493.1 | |
NM_002669.3 | 1578 | Missense Mutation | ACA,CCA | T,P 494 | NP_002660.1 |