Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCACATGGCAAATGGGATTGAAGA[C/T]TGTGATGATCAGCCACCTAAAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604215 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ING2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ING2 - inhibitor of growth family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291959.1 | 507 | Silent Mutation | GAC,GAT | D,D 132 | NP_001278888.1 | |
NM_001564.3 | 507 | Silent Mutation | GAC,GAT | D,D 172 | NP_001555.1 | |
XM_011531927.2 | 507 | Silent Mutation | GAC,GAT | D,D 117 | XP_011530229.1 |
LOC389247 - uncharacterized LOC389247 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |