Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATAATGGTGTTTCCTTTTTATAG[A/T]TGAAGCTAGGGTCAGAACCTTCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD50 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD50 - ankyrin repeat domain 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167882.1 | 6036 | Missense Mutation | AAC,ATC | N,I 1243 | NP_001161354.1 | |
NM_020337.2 | 6036 | Missense Mutation | AAC,ATC | N,I 1422 | NP_065070.1 | |
XM_017008471.1 | 6036 | Missense Mutation | AAC,ATC | N,I 1422 | XP_016863960.1 |