Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACTCAGGGTGCCGGTACTTATCC[C/T]GCGCCTTCACCACAGCACCGCTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605889 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDLIM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDLIM3 - PDZ and LIM domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114107.4 | 906 | Missense Mutation | CAG,CGG | Q,R 261 | NP_001107579.1 | |
NM_001257962.1 | 906 | Missense Mutation | CAG,CGG | Q,R 221 | NP_001244891.1 | |
NM_001257963.1 | 906 | Missense Mutation | CAG,CGG | Q,R 142 | NP_001244892.1 | |
NM_014476.5 | 906 | Missense Mutation | CAG,CGG | Q,R 309 | NP_055291.2 |