Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATACCACCAGCCTCCCCCATAGAC[C/T]TCTGCACAGTTCTCTTCCCACTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 134820 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGA - fibrinogen alpha chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000508.4 | 2482 | Silent Mutation | GAA,GAG | E,E 801 | NP_000499.1 | |
NM_021871.3 | 2482 | Intron | NP_068657.1 |