Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTTTATCAAATTTTTCATTCAGG[A/G]CTACTGTAAAGTGATTTTTCCAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600043 | ||||||||||||||||||||
Literature Links: |
SULT1E1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SULT1E1 - sulfotransferase family 1E member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005420.2 | 922 | Missense Mutation | GCC,GTC | A,V 270 | NP_005411.1 | |
XM_011532210.2 | 922 | Intron | XP_011530512.1 |