Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCAGAAGGCATTTCATAAGCCT[C/T]ATTGTCAGGATCCACAGGCATATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 163890 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNCA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNCA - synuclein alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000345.3 | 437 | Missense Mutation | AAG,GAG | K,E 123 | NP_000336.1 | |
NM_001146054.1 | 437 | Missense Mutation | AAG,GAG | K,E 123 | NP_001139526.1 | |
NM_001146055.1 | 437 | Missense Mutation | AAG,GAG | K,E 123 | NP_001139527.1 | |
NM_007308.2 | 437 | Intron | NP_009292.1 | |||
XM_011532203.1 | 437 | Intron | XP_011530505.1 | |||
XM_011532204.2 | 437 | Intron | XP_011530506.1 | |||
XM_011532205.2 | 437 | Intron | XP_011530507.1 | |||
XM_011532206.1 | 437 | Intron | XP_011530508.1 | |||
XM_011532207.1 | 437 | Intron | XP_011530509.1 | |||
XM_011532208.2 | 437 | Missense Mutation | AAG,GAG | K,E 123 | XP_011530510.1 | |
XM_017008562.1 | 437 | Missense Mutation | AAG,GAG | K,E 123 | XP_016864051.1 | |
XM_017008563.1 | 437 | Missense Mutation | AAG,GAG | K,E 123 | XP_016864052.1 |